missing translation for 'onlineSavingsMsg'
Learn More

SPDB-DM4, MedChemExpress

Product Code. 16455178
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16455178 10 mg 10mg
16441259 1 mg 1mg
16407907 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16455178 Supplier Medchem Express Supplier No. HY12460
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SPDB-DM4 is a drug-linker conjugate for ADC by using the maytansinebased payload (DM4, a tubulin inhibitor) via a SPDB linker, exhibiting potent anti-tumor activity.

SPDB-DM4 is a drug-linker conjugate for ADC by using the maytansinebased payload (DM4, a tubulin inhibitor) via a SPDB linker, exhibiting potent anti-tumor activity.

Specifications

Chemical Name or Material SPDB-DM4
CAS 1626359-62-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C46H63ClN4O14S2
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (50.22 mM; Need ultrasonic)
SMILES C[C@]1([C@H](CC(N(C(C=C2C=C3OC)=C3Cl)C)=O)OC([C@H](C)N(C)C(CCC(C)(C)SSCCCC(ON(C4=O)C(CC4)=O)=O)=O)=O)[C@H]([C@@H]([C@](OC5=O)([H])C[C@]([C@](/C=C/C=C(C)/C2)([H])OC)(N5)O)C)O1
Molecular Weight (g/mol) 995.59
Formula Weight 995.59
Percent Purity 95.48%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.