missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SPCS2 Polyclonal antibody specifically detects SPCS2 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
Specifications
| Antigen | SPCS2 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | MGC117366, signal peptidase complex subunit 2, signal peptidase complex subunit 2 homolog (S. cerevisiae), SPC25 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human SPCS2 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?