missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPC25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561444
This item is not returnable.
View return policy
Description
Immunogen sequence: RLGLEIRKIY GEKLQFIFTN IDPKNPESPF MFSLHLNEAR DYEVSDSAPH LEGLAEFQEN VRKTNNFSAF LANVRKAFTA TVY Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 83%.
This gene encodes a protein that may be involved in kinetochore-microtubule interaction and spindle checkpoint activity.
Specifications
| SPC25 | |
| Polyclonal | |
| Unconjugated | |
| SPC25 | |
| 2600017H08Rik; 2610205L13Rik; AD024; hSpc25; kinetochore protein Spc25; RGD1307811; SPBC25; SPC25; SPC25, NDC80 kinetochore complex component; SPC25, NDC80 kinetochore complex component, homolog; SPC25, NDC80 kinetochore complex component, homolog (S. cerevisiae); spindle pole body component 25 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57405 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HBM1 | |
| SPC25 | |
| Recombinant protein corresponding to Human SPC25. Recombinant protein control fragment (Product #RP-98498). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction