missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPC24 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97666-100ul
This item is not returnable.
View return policy
Description
SPC24 Polyclonal antibody specifically detects SPC24 in Human samples. It is validated for Western Blot
Specifications
| SPC24 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ90806, hSpc24, kinetochore protein Spc24, SPBC24, SPC24, NDC80 kinetochore complex component, homolog (S. cerevisiae), spindle pole body component 24 homolog, spindle pole body component 24 homolog (S. cerevisiae) | |
| Produced in rabbits immunized with E. coli-derived Human SPC24 fragment. | |
| 100 μg | |
| Apoptosis, Cell Biology, Cell Cycle and Replication | |
| 147841 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction