missing translation for 'onlineSavingsMsg'
Learn More

SPC24 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18405677 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405677 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405677 Supplier Novus Biologicals Supplier No. NBP232105AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPC24 Polyclonal antibody specifically detects SPC24 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPC24
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FLJ90806, hSpc24, kinetochore protein Spc24, SPBC24, SPC24, NDC80 kinetochore complex component, homolog (S. cerevisiae), spindle pole body component 24 homolog, spindle pole body component 24 homolog (S. cerevisiae)
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Spindle Pole Body Component 24 using the numbering given in entry NP_872319.1 (GeneID 147841).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 147841
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.