missing translation for 'onlineSavingsMsg'
Learn More

SPB, MedChemExpress

Product Code. 16473928
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473928 10 mg 10mg
16456647 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16473928 Supplier Medchem Express Supplier No. HY104025
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SPB is a drug-linker conjugate for ADC with potent anti-inflammatory activity by using Xanthotoxol, linked via the ADC linker.

SPB is a drug-linker conjugate for ADC with potent anti-inflammatory activity by using Xanthotoxol, linked via the ADC linker.

Specifications

Chemical Name or Material SPB
CAS 858128-57-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H15NO8
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (129.76 mM; Need ultrasonic)
SMILES O=C(ON1C(CCC1=O)=O)CCCOC2=C(O3)C(C=CC3=O)=CC4=C2OC=C4
Molecular Weight (g/mol) 385.32
Formula Weight 385.32
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.