missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPATS2L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566836
This item is not returnable.
View return policy
Description
Immunogen sequence: GEITHPKNNYS SRTPCSSLLP LLNAHAATSG KQSNFSRKSS THNKPSEGKA ANPKMVSSLP STADPSHQTM PANKQNGSSN QRRRFN Highest antigen sequence identity to the following orthologs - mouse 83%, rat 85%.
SPATS2L is a protein coding gene.
Specifications
| SPATS2L | |
| Polyclonal | |
| Unconjugated | |
| SPATS2L | |
| 2810022L02Rik; A230104H11Rik; AA987173; AI480531; AI842453; AW413586; DNA polymerase transactivated protein 6; DNA polymerase-transactivated protein 6; DNAPTP6; RGD1309930; SGNP; SP1224; Spats2l; SPATS2-like protein; spermatogenesis associated serine rich 2 like; spermatogenesis associated, serine rich 2 like; spermatogenesis associated, serine-rich 2-like; stress granule and nucleolar protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26010 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NUQ6 | |
| SPATS2L | |
| Recombinant Human SPATS2L. Recombinant protein control fragment (Product #RP-105390). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction