missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPATS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558477
This item is not returnable.
View return policy
Description
Immunogen sequence: SKSVSIQEEQ SAPSSEKGGM NGYHVNGAIN DTESVDSLSE GLETLSIDAR ELEDPESAML DTLDRTGSML QNGVSDFETK SLTMHSIHNS QQPRNAAKSL SRPTTETQ Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 78%.
SPATS2 (Spermatogenesis Associated Serine Rich 2) is a Protein Coding gene. Diseases associated with SPATS2 include Progesterone-Receptor Negative Breast Cancer. An important paralog of this gene is SPATS2L. [GeneCards].
Specifications
| SPATS2 | |
| Polyclonal | |
| Unconjugated | |
| SPATS2 | |
| Nbla00526; P59SCR; putative protein product of Nbla00526; SCR59; Serine-rich spermatocytes and round spermatid 59 kDa protein; serine-rich spermatocytes and round spermatid protein; SPATA10; SPATS2; spermatogenesis associated serine rich 2; spermatogenesis associated, serine rich 2; spermatogenesis associated, serine-rich 2; spermatogenesis-associated serine-rich protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 65244 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86XZ4 | |
| SPATS2 | |
| Recombinant protein corresponding to Human SPATS2. Recombinant protein control fragment (Product #RP-97722). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction