Learn More
Invitrogen™ SPATS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110419
Description
Immunogen sequence: MSRKQNQKDS SGFIFDLQSN TVLAQGGAFE NMKEKINAVR AIVPNKSNNE IILVLQHFDN CVDKTVQAFM EGSASEVLKE WTVTGKKKNK KKKNKPKPAA EPSNGIPDSS KSVSIQEEQS APSSEKGGMN GYHVNGAIND TESVDSLSEG LETLSIDARE LEDPESAMLD TLDRTGSMLQ NGVSDFETKS LTMHSIHNSQ QPRNAAKSLS RPTTETQFSN MGMEDVPLAT SKKLSSNIEK.
SPATS2 (Spermatogenesis Associated Serine Rich 2) is a Protein Coding gene. Diseases associated with SPATS2 include Progesterone-Receptor Negative Breast Cancer. An important paralog of this gene is SPATS2L. [GeneCards].
Specifications
| SPATS2 | |
| Polyclonal | |
| Unconjugated | |
| SPATS2 | |
| Nbla00526; P59SCR; putative protein product of Nbla00526; SCR59; Serine-rich spermatocytes and round spermatid 59 kDa protein; serine-rich spermatocytes and round spermatid protein; SPATA10; SPATS2; spermatogenesis associated serine rich 2; spermatogenesis associated, serine rich 2; spermatogenesis associated, serine-rich 2; spermatogenesis-associated serine-rich protein 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 300221 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| 0 | |
| SPATS2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human SPATS2 (NP_075559.2). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.