missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97797-100ul
This item is not returnable.
View return policy
Description
SPATS2 Polyclonal antibody specifically detects SPATS2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SPATS2 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| FLJ13117, P59SCR, SCR59putative protein product of Nbla00526, Serine-rich spermatocytes and round spermatid 59 kDa protein, SPATA10serine-rich spermatocytes and round spermatid protein, spermatogenesis associated, serine-rich 2, spermatogenesis-associated serine-rich protein 2 | |
| Produced in rabbits immunized with E. coli-derived Human SPATS2 fragment. | |
| 100 μg | |
| Cell Biology | |
| 65244 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction