missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | SPATC1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SPATC1 Polyclonal specifically detects SPATC1 in Human samples. It is validated for Western Blot.Specifications
| SPATC1 | |
| Polyclonal | |
| Rabbit | |
| MGC61633, SPATA15SPERIOLIN, speriolin, spermatogenesis and centriole associated 1, Spermatogenesis and centriole-associated protein 1, Spermatogenesis-associated protein 15, Spermatogenic cell-specific Cdc20-binding protein, SPRN | |
| SPATC1 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 375686 | |
| Synthetic peptides corresponding to SPATC1(spermatogenesis and centriole associated 1) The peptide sequence was selected from the middle region of SPATC1. Peptide sequence QSSPLIAPVMGTVAVSLSSPLLSSTATPPGVSQNLLANPMSNLVLPEAPR. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title