missing translation for 'onlineSavingsMsg'
Learn More

SPATA6 Antibody - BSA Free, Novus Biologicals™

Product Code. 18365970 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365970 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18365970 Supplier Novus Biologicals Supplier No. NBP281853

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPATA6 Polyclonal specifically detects SPATA6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA6
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias HASH, spermatogenesis associated 6
Gene Symbols SPATA6
Host Species Rabbit
Immunogen SPATA6 antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human SPATA6. The immunogen is located within amino acids 400 - 450 of SPATA6. Amino Acid Squence: RDSAYDSDPEYSS
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54558
Test Specificity At least three isoforms of SPATA6 are known to exist; this antibody will detect only the longest two isoforms. SPATA6 antibody is predicted to not cross-react with other SPATA protein family members.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.