missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ SPATA5L1 Monoclonal Antibody (16A2)

SPATA5L1 Monoclonal Antibody (16A2)
Brand: Thermo Scientific™ MA534952
This item is not returnable.
View return policy
Description
SPATA5L1 Monoclonal Antibody for ICC/IF, IHC (P), Flow
Specifications
| SPATA5L1 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9BVQ7 | |
| SPATA5L1 | |
| Recombinant protein within Human SPATA5L1 aa 379-580. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 16A2 | |
| Unconjugated | |
| SPATA5L1 | |
| AV141009; C130039A10Rik; SPATA5L1; spermatogenesis associated 5 like 1; spermatogenesis associated 5-like 1; spermatogenesis-associated protein 5-like protein 1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 79029 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction