missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | SPATA5 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18351857
|
Novus Biologicals
NBP3-16028-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18347323
|
Novus Biologicals
NBP3-16028-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPATA5 Polyclonal antibody specifically detects SPATA5 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SPATA5 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, 50% glycerol, pH7.3 | |
| 166378 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| AFG2ATPase family gene 2 homolog, ATPase family protein 2 homolog, SPAFspermatogenesis associated factor SPAF, spermatogenesis associated 5, Spermatogenesis-associated factor protein, spermatogenesis-associated protein 5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 228-304 of human SPATA5 (NP_001304728). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title