Learn More
Invitrogen™ SPATA4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596732
Description
Immunogen sequence: MAAAGQEKGY LTQTAAALDK SPSLSPQLAA PIRGRPKKCL VYPHAPKSSR LSRSVLRWLQ GLDLSFFPRN INRDFSNGFL IAEIFCIYYP WELELSSFEN GTSLKVKLDN WAQLEKFLAR KKFKLPKELI HGTIHCKAGV PEILIEEVYT LLTHREIKSI QDDFVNFTDY SYQMRLPLVS RSTVSKSIKD NIRLSELLSN PNMLTNELKA EFLILLHMLQ RKLGRKLNPE WFDVKPTVGE VTLNHLPAQA; Positive Samples: Mouse heart, Rat heart.
The testis spermatocyte apoptosis-related gene 2 protein (TSARG2, also designated spermatogenesis associated-4 or SPATA4) is involved in spermatogenesis. TSARG2 is specifically expressed in spermatogonia and spermatocytes of the seminiferous tubules, and it localizes to the nucleus. The predicted molecular weight of TSARG2 ranges depending on the species. TSARG2 is significantly upregulated in cryptorchidism and therefore, is a testis-specific apoptosis candidate oncogene.
Specifications
| SPATA4 | |
| Polyclonal | |
| Unconjugated | |
| SPATA4 | |
| 1700001N01Rik; Spata4; SPEF1B; spermatogenesis associated 4; spermatogenesis related gene 2; spermatogenesis-associated 4; spermatogenesis-associated protein 4; Spermatogenesis-related gene 2 protein; Srg2; testis and spermatogenesis cell related protein 2; testis and spermatogenesis cell-related protein 2; Testis spermatocyte apoptosis-related gene 2 protein; TSARG2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 132851, 306441, 69281 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.57 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8K3V1, Q8NEY3 | |
| SPATA4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human SPATA4 (NP_6532452). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.