missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPATA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572724
This item is not returnable.
View return policy
Description
SPATA4 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The testis spermatocyte apoptosis-related gene 2 protein (TSARG2, also designated spermatogenesis associated-4 or SPATA4) is involved in spermatogenesis. TSARG2 is specifically expressed in spermatogonia and spermatocytes of the seminiferous tubules, and it localizes to the nucleus. The predicted molecular weight of TSARG2 ranges depending on the species. TSARG2 is significantly upregulated in cryptorchidism and therefore, is a testis-specific apoptosis candidate oncogene.
Specifications
| SPATA4 | |
| Polyclonal | |
| Unconjugated | |
| SPATA4 | |
| 1700001N01Rik; Spata4; SPEF1B; spermatogenesis associated 4; spermatogenesis related gene 2; spermatogenesis-associated 4; spermatogenesis-associated protein 4; Spermatogenesis-related gene 2 protein; Srg2; testis and spermatogenesis cell related protein 2; testis and spermatogenesis cell-related protein 2; Testis spermatocyte apoptosis-related gene 2 protein; TSARG2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 132851, 69281 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8K3V1, Q8NEY3 | |
| SPATA4 | |
| SPATA4 antibody for an 18 amino acid synthetic peptide near the carboxy terminus of human SPATA4. The immunogen is within amino acids 200 - 250 of SPATA4. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction