missing translation for 'onlineSavingsMsg'
Learn More

SPATA4 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18415545 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415545 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415545 Supplier Novus Biologicals Supplier No. NBP281852JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPATA4 Polyclonal antibody specifically detects SPATA4 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA4
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias SPEF1B, spermatogenesis associated 4, spermatogenesis-associated protein 4, testis and spermatogenesis cell related protein 2, Testis and spermatogenesis cell-related protein 2, Testis spermatocyte apoptosis-related gene 2 protein, TSARG2MGC33432
Host Species Rabbit
Immunogen SPATA4 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human SPATA4. The immunogen is located within amino acids 200 - 250 of SPATA4. Amino Acid Squence: LHMLQRKLGRKLNPEWFD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 132851
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.