missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93377-0.02ml
This item is not returnable.
View return policy
Description
SPATA4 Polyclonal antibody specifically detects SPATA4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SPATA4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| SPEF1B, spermatogenesis associated 4, spermatogenesis-associated protein 4, testis and spermatogenesis cell related protein 2, Testis and spermatogenesis cell-related protein 2, Testis spermatocyte apoptosis-related gene 2 protein, TSARG2MGC33432 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human SPATA4 (NP_653245.2). MAAAGQEKGYLTQTAAALDKSPSLSPQLAAPIRGRPKKCLVYPHAPKSSRLSRSVLRWLQGLDLSFFPRNINRDFSNGFLIAEIFCIYYPWELELSSFENGTSLKVKLDNWAQLEKFLARKKFKLPKELIHGTIHCKAGVPEILIEEVYTLLTHREIKSIQDDFVNFTDYSYQMRLPLVSRSTVSKSIKDNIRLSELLSNPNMLTNELKAEFLILLHMLQRKLGRKLNPEWFDVKPTVGEVTLNHLPAQA | |
| 0.02 mL | |
| Apoptosis | |
| 132851 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction