missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SPATA31E1 Polyclonal specifically detects SPATA31E1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | SPATA31E1 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 405 |
| Dilution | Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C9orf79, chromosome 9 open reading frame 79, FAM75E1, FAM75-like protein C9orf79, SPATA31 subfamily E, member 1, XXyac-YM21GA2.5 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human SPATA31E1 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?