missing translation for 'onlineSavingsMsg'
Learn More

SPATA3 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18419307 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419307 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419307 Supplier Novus Biologicals Supplier No. NBP281816V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPATA3 Polyclonal antibody specifically detects SPATA3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA3
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias spermatogenesis associated 3, spermatogenesis-associated protein 3, testis and spermatogenesis cell apoptosis related gene 1, Testis spermatocyte apoptosis-related protein 1, TSARG1Testis and spermatogenesis cell-related protein 1
Host Species Rabbit
Immunogen SPATA3 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human SPATA3. The immunogen is located within the last 50 amino acids of SPATA3. Amino Acid Squence: LTFYRKSSRKPSSHRNA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 130560
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.