missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA22 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16989-1-AP-20UL
This item is not returnable.
View return policy
Description
SPATA22 is highly expressed in adult testis. The specific function of SPATA22 is not yet known.Specifications
| SPATA22 | |
| Polyclonal | |
| Unconjugated | |
| SPATA22 | |
| NYD SP20, SPATA22, spermatogenesis associated 22 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 380709, 84690 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5SV06, Q8NHS9 | |
| Spata22 | |
| SPATA22 Fusion Protein Ag10591 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction