missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA22 Antibody (OTI6H9), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74309C
This item is not returnable.
View return policy
Description
SPATA22 Monoclonal specifically detects SPATA22 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SPATA22 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SPATA22 | |
| Full length recombinant protein of human SPATA22 (NP_115987) produced in E.coli. | |
| 0.1 mL | |
| 84690.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI6H9 | |
| Western Blot, Immunohistochemistry | |
| NYDSP20, NYD-SP20, spermatogenesis associated 22, spermatogenesis-associated protein 22, testes development-related NYD-SP20, Testis development protein NYD-SP20 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction