missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SPATA22 Monoclonal antibody specifically detects SPATA22 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
Specifications
| Antigen | SPATA22 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI6H9 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | NYDSP20, NYD-SP20, spermatogenesis associated 22, spermatogenesis-associated protein 22, testes development-related NYD-SP20, Testis development protein NYD-SP20 |
| Host Species | Mouse |
| Immunogen | Full length recombinant protein of human SPATA22 (NP_115987) produced in E.coli. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?