missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPATA20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550575
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total SPATA20 protein.
The protein may play a role in fertility regulation. It were significantly upregulated in individual bile samples from Cholangiocarcinoma patients by Western blotting and Immunohistochemistry, relative to normal tissues. It acts as a potential serum diagnostic biomarker for Cholangiocarcinoma.
Specifications
| SPATA20 | |
| Polyclonal | |
| Unconjugated | |
| SPATA20 | |
| epididymis secretory protein Li 98; HEL-S-98; Spata20; sperm protein SSP411; spermatogenesis associated 20; spermatogenesis-associated protein 20; sperm-specific protein 411; SSP411; Tisp78; transcript increased in spermiogenesis 78; transcript increased in spermiogenesis 78 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64847 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2.8 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8TB22 | |
| SPATA20 | |
| Synthetic peptide corresponding to a region derived from internal residues of human spermatogenesis associated 20. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction