missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA20 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£342.00 - £571.00
Specifications
| Antigen | SPATA20 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18412431
|
Novus Biologicals
NBP1-82147 |
0.1 mL |
£571.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18421521
|
Novus Biologicals
NBP1-82147-25ul |
25 μL |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPATA20 Polyclonal antibody specifically detects SPATA20 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| SPATA20 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp686H1839, FLJ21347, FLJ21969, MGC111032, sperm protein SSP411, spermatogenesis associated 20, spermatogenesis-associated protein 20, Sperm-specific protein 411, Ssp411, Tisp78, transcript increased in spermiogenesis 78 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SHPSSTPQRVPNRLIHEKSPYLLQHAYNPVDWYPWGQEAFDKARKENKPIFLSVGYSTCHWCHMMEEESFQNEEIGRLLSEDFVSVKV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 ug/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 64847 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title