Learn More
SPATA2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29321-1-AP-20UL
Description
The SPATA2 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human SPATA2. This antibody recognizes human, mouse, rat antigen. The SPATA2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| SPATA2 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| 20 μL | |
| 9825 | |
| IgG |