missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPATA16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583394
This item is not returnable.
View return policy
Description
Immunogen sequence: LQALHMLPQT VDWSSFPPQQ YLLTLGFKNK DDGKFLEKIS SRKLPIFTEH KTPFGLTRED TVRQMETMGK RIL Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 82%.
This gene encodes a testis-specific protein that belongs to the tetratricopeptide repeat-like superfamily. The encoded protein localizes to the Golgi apparatus and may play a role in spermatogenesis.
Specifications
| SPATA16 | |
| Polyclonal | |
| Unconjugated | |
| SPATA16 | |
| 4921511F01Rik; 4930503K02Rik; CDC68; FACT; FACTP140; FLJ10857; FLJ14010; NYD-SP12; RGD1563726; Spata16; spermatogenesis associated 16; spermatogenesis-associated protein 16; spermatogenesis-related protein; SPGF6; SUPT16H; testicular tissue protein Li 185; testis development protein NYD-SP12; testis-specific Golgi protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83893 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BXB7 | |
| SPATA16 | |
| Recombinant protein corresponding to Human SPATA16. Recombinant protein control fragment (Product #RP-96368). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction