missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPATA16 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11546072
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11546072 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11546072 Supplier Invitrogen™ Supplier No. PA112696

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPATA16 Polyclonal Antibody for Western Blot, ChIP

This gene encodes a testis-specific protein that belongs to the tetratricopeptide repeat-like superfamily. The encoded protein localizes to the Golgi apparatus and may play a role in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA16
Applications ChIP Assay, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene SPATA16
Gene Accession No. Q8C636, Q9BXB7
Gene Alias 4921511F01Rik; 4930503K02Rik; CDC68; FACT; FACTP140; FLJ10857; FLJ14010; NYD-SP12; RGD1563726; Spata16; spermatogenesis associated 16; spermatogenesis-associated protein 16; spermatogenesis-related protein; SPGF6; SUPT16H; testicular tissue protein Li 185; testis development protein NYD-SP12; testis-specific Golgi protein
Gene Symbols SPATA16
Host Species Rabbit
Immunogen Recombinant protein consisting of amino acids 1-329 of the human SPT16 protein conjugated to KLH.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 70862, 83893
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.