Learn More
Invitrogen™ SPATA12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139995
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: TQHPNKPHCA LASCQGPGVL PGAASALPEL TFQGDVCQSE TCQRYLQAAI SLDIA Highest antigen sequence indentity to the following orthologs: Rat - 34%, Mouse - 34%.
This gene is expressed primarily in testis and may play a role in testicular development and spermatogenesis. The encoded protein may be upregulated in response to ultraviolet-C radiation. [provided by RefSeq, Dec 2015].
Specifications
| SPATA12 | |
| Polyclonal | |
| Unconjugated | |
| SPATA12 | |
| SPATA12; spermatogenesis associated 12; spermatogenesis-associated protein 12; Spermatogenesis-related protein 5; SRG5; testicular tissue protein Li 184 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 353324 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z6I5 | |
| SPATA12 | |
| Recombinant protein corresponding to Human SPATA12. Recombinant protein control fragment (Product #RP-109489). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.