missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPAT5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557664
This item is not returnable.
View return policy
Description
Immunogen sequence: SLELSLQLSQ LDLEDTQIPT SRSTPYKPID DRITNKASDV LLDVTQSPGD GSGLMLEEVT GLKCNFESAR EGNEQLT Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 65%.
This gene encodes a member of the ATPase associated with diverse activities family, whose members are defined by a highly conserved ATPase domain. Members of this family participate in diverse cellular processes that include membrane fusion, DNA replication, microtubule severing, and protein degradation. The protein encoded by this gene has a putative mitochondrial targeting sequence and has been proposed to function in maintenance of mitochondrial function and integrity during mouse spermatogenesis. Allelic variants in this gene have been associated with epilepsy, hearing loss, and cognitive disability syndrome. Alternative splicing results in multiple transcript variants.
Specifications
| SPAT5 | |
| Polyclonal | |
| Unconjugated | |
| SPATA5 | |
| 2510048F20Rik; AFG2; ATPase family gene 2 homolog; ATPase family protein 2 homolog; C78064; EHLMRS; Spaf; Spata5; spermatogenesis associated 5; spermatogenesis associated factor; spermatogenesis associated factor SPAF; spermatogenesis-associated factor protein; Spermatogenesis-associated protein 5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 166378 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NB90 | |
| SPATA5 | |
| Recombinant protein corresponding to Human SPAT5. Recombinant protein control fragment (Product #RP-102718). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction