missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Spastin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544807
This item is not returnable.
View return policy
Description
Peptide sequence: RVLVMGATNR PQELDEAVLR RFIKRVYVSL PNEETRLLLL KNLLCKQGSP Sequence homology: Cow: 100%; Dog: 100%; Goat: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
This gene encodes a member of the AAA (ATPases associated with a variety of cellular activities) protein family. Members of this protein family share an ATPase domain and have roles in diverse cellular processes including membrane trafficking, intracellular motility, organelle biogenesis, protein folding, and proteolysis. The encoded ATPase may be involved in the assembly or function of nuclear protein complexes. Two transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants have been described but their full length sequences have not been determined. Mutations associated with this gene cause the most frequent form of autosomal dominant spastic paraplegia 4.
Specifications
| Spastin | |
| Polyclonal | |
| Unconjugated | |
| SPAST | |
| ADPSP; ATPase; fidgetin-like 1; FSP2; HAMAP-Rule:MF_03021}; hypothetical protein zgc:85952; Kiaa1083; KIAA183; LOW QUALITY PROTEIN: spastin; mKIAA1083; si:ch73-233a22.3; SPAST; spastic paraplegia 4 (autosomal dominant; spastic paraplegia 4 (autosomal dominant; spastin); spastic paraplegia 4 homolog; spastic paraplegia 4 protein; Spastin; spastin {ECO:0000255; SPG4; zgc:85952 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6683 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UBP0 | |
| SPAST | |
| Synthetic peptide directed towards the middle region of human SPAST. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction