missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spastin Antibody (Sp 6C6), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50303JF549
This item is not returnable.
View return policy
Description
Spastin Monoclonal antibody specifically detects Spastin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Spastin | |
| Monoclonal | |
| Janelia Fluor 549 | |
| ADPSPSpastic paraplegia 4 protein, FSP2EC 3.6.4.3, KIAA1083EC 3.6.1.3, spastic paraplegia 4 (autosomal dominant, spastin), spastin, SPG4 | |
| Recombinant Spastin | |
| 0.1 mL | |
| Cardiovascular Biology, Cell Cycle and Replication, Cellular Signaling, Epigenetics, metabolism, Neuroscience | |
| 6683 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, ELISA, Immunofluorescence | |
| Sp 6C6 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction