missing translation for 'onlineSavingsMsg'
Learn More

Sparsentan, MedChemExpress

Product Code. 16447428
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16447428 10 mg 10mg
16439907 5 mg 5mg
16427406 100 mg 100mg
16423586 25 mg 25mg
16413457 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16447428 Supplier Medchem Express Supplier No. HY17621
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sparsentan (RE-021) is a highly potent dual angiotensin II and endothelin A receptor antagonist with Kis of 0.8 and 9.3 nM, respectively.

Sparsentan (RE-021) is a highly potent dual angiotensin II and endothelin A receptor antagonist with Kis of 0.8 and 9.3 nM, respectively.

Specifications

Chemical Name or Material Sparsentan
CAS 254740-64-2
Color White
Purity Grade Notes Research
Molecular Formula C32H40N4O5S
Quantity 10 mg
Synonym RE-021 DARA-a
Solubility Information DMSO : 100 mg/mL (168.71 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC=C1C2=CC=C(CN3C(CCCC)=NC4(CCCC4)C3=O)C=C2COCC)(NC5=NOC(C)=C5C)=O
Molecular Weight (g/mol) 592.75
Formula Weight 592.75
Percent Purity 98.01%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.