missing translation for 'onlineSavingsMsg'
Learn More

Sparfloxacin, MedChemExpress

Product Code. p-200097466
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16478226

Brand: Medchem Express HYB0308

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sparfloxacin (CI-978) is a fluoroquinolone antibiotic, shows broad and potent antibacterial activity.

Sparfloxacin (CI-978) is a fluoroquinolone antibiotic, shows broad and potent antibacterial activity.

Specifications

Chemical Name or Material Sparfloxacin
Physical Form Powder
CAS 110871-86-8
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H22F2N4O3
For Use With (Application) COVID-19-immunoregulation
Synonym CI-978 AT-4140
Solubility Information 0.1 M NaOH : 50 mg/mL (127.42 mM; ultrasonic and adjust pH to 11 with NaOH) ∣DMSO : 3.33 mg/mL (8.49 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CN(C2CC2)C3=C(C(N)=C(F)C(N4C[C@H](C)N[C@H](C)C4)=C3F)C1=O)O
Molecular Weight (g/mol) 392.4
Formula Weight 392.4
Percent Purity 98.07%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.