Learn More
Biochrom Spare Power Supply for Ultrospec 1100 UV-Vis Spectrophotometer
Click to view available options
For Use With (Equipment):
Biochrom Ultrospec 1100 UV-Vis Spectrophotometer
Unit Size:
Each
Description
- Ready to use
Specifications
Specifications
| Product Type | Spare Part |
| For Use With (Equipment) | Biochrom Ultrospec 1100 UV-Vis Spectrophotometer |
| Material | Polypropylene |
| Voltage | 18 to 32 VDC |
| Autoclavable | Not Autoclavable |
| Description | Spare Power Supply for Ultrospec 1100 UV-Vis Spectrophotometer |