missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Spare Plunger

Product Code. 10238340
Click to view available options
Volume (Metric):
5 to 25 μL
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 10238340

Brand: Socorex™ 342.025

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Order Info

dimColor

Specifications

Volume (Metric) 5 to 25 μL
Color White
For Use With (Application) For Acura 846 Positive Displacement Pipettes
Quantity 5 Pack
Product Type Spare Plunger
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.