Learn More
Brand™ Spare Key for Intermediate Stopcock 4 NS/19
For use with automatic burettes, pellet pattern.
Brand: Brand™ 81256
Additional Details : Weight : 0.04000kg
Description
- Manufactured from borosilicate glass 5.4, either clear or amber tint
- With retention device
- Provided individually
Specifications
Borosilicate Glass | |
1/Pk. | |
Retention device | |
Amber |