Learn More
Invitrogen™ SPARC Monoclonal Antibody (ON1-1)

Description
Source: Monoclonal antibody was obtained by fusing the mouse myeloma cell-line P3-X63-Ag8-U1 with spleen cells of BALB/c mouse after immunization with bovine bone osteonectin (SPARC). The monoclonal antibody was harvested from ascitic fluid. Specificity: This antibody reacts with both bone derived and platelet derived Osteonectin (ELISA). This antibody does not react with non-activated platelet and reacts with thrombin-stimulated platelet (flow cytometric analysis). This antibody reacts with human and bovine Osteonectin (Western blot analysis). This antibody cross reacts with rabbit and porcine Osteonectin. (ELISA) Storage: 4°C. This product does not contain preservative. The stock solution (2.0 mg/mL) should be stored in aliquots at -20°C for 1 year, or should be stored at 4°C for 6 months after adding 0.1% sodium azide. Avoid repeated freeze-thaw cycles. Reconstitution: Dissolve the lyophilized antibody in 50 μL of distilled water (final concentration: 2.0 mg/mL). This solution can be used as a stock solution. If dilution is necessary for your application, dilute the stock solution with the following Dilution solution just prior to use. When the entire amount of antibody is to be used over a short time period, it may be dissolved directly in 500 μL or more of the Dilution solution. Be sure to store the antibody at a minimum concentration of 2.0 mg/mL. A lower antibody concentration may result in decreased stability. Reconstituted antibody solution shoul...
Specifications
Specifications
| Antigen | SPARC |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | ON1-1 |
| Concentration | 2.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA and no preservative; pH 7.4 |
| Gene | SPARC |
| Gene Accession No. | P09486, P13213, P20112, P36233 |
| Gene Alias | Basement membrane protein 40; basement-membrane protein 40; BM40; BM-40; Cysteine rich protein; OI17; ON; Osteonectin; osteonectin precursor; Secreted acidic cystein rich glycoprotein; secreted acidic cysteine rich glycoprotein; Secreted acidic cystein-rich glycoprotein (osteonectin); secreted protein acidic and cysteine rich; secreted protein acidic and rich in cysteine; Secreted protein acidic and rich in cysteine (SPARC); secreted protein, acidic, cysteine-rich (osteonectin); secreted protein, acidic, cysteine-rich [osteonectin]; SPARC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.