missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPARC-like 1/SPARCL1 Antibody Pair (HRP), Novus Biologicals™
Mouse Monoclonal Antibody
£292.00
Specifications
| Antigen | SPARC-like 1/SPARCL1 |
|---|---|
| Dilution | Sandwich ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Host Species | Mouse |
Description
SPARC-like 1/SPARCL1 Monoclonal specifically detects SPARC-like 1/SPARCL1 in Human samples. It is validated for ELISA, Sandwich ELISA.Specifications
| SPARC-like 1/SPARCL1 | |
| Monoclonal | |
| Mouse | |
| 8404 | |
| Storage of components varies. See protocol for specific instructions. |
| Sandwich ELISA | |
| HRP | |
| hevin, High endothelial venule protein, MAST 9, MAST9, PIG33, proliferation-inducing protein 33, SC1, SPARC-like 1 (hevin), SPARC-like 1 (mast9, hevin), SPARC-like protein 1 | |
| SPARCL1 | |
| This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human SPARC-like 1/SPARCL1. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title