missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPARC-like 1/SPARCL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £255.00
Specifications
| Antigen | SPARC-like 1/SPARCL1 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18640602
|
Novus Biologicals
NBP2-98296-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18607884
|
Novus Biologicals
NBP2-98296-50ul |
50 μg |
£178.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPARC-like 1/SPARCL1 Polyclonal antibody specifically detects SPARC-like 1/SPARCL1 in Mouse samples. It is validated for ELISASpecifications
| SPARC-like 1/SPARCL1 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 0.2 um filtered solution in PBS | |
| 8404 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| hevin, High endothelial venule protein, MAST 9, MAST9, PIG33, proliferation-inducing protein 33, SC1, SPARC-like 1 (hevin), SPARC-like 1 (mast9, hevin), SPARC-like protein 1 | |
| Produced in rabbits immunized with purified, recombinant Mouse SPARC-like 1/SPARCL1 (Accession#: EDL20231.1; Met1-Phe650) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title