missing translation for 'onlineSavingsMsg'
Learn More

SPARC-like 1/SPARCL1 Antibody (390), DyLight 650, Novus Biologicals™

Product Code. 18323391 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323391 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18323391 Supplier Novus Biologicals Supplier No. NBP289237C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SPARC-like 1/SPARCL1 Monoclonal specifically detects SPARC-like 1/SPARCL1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPARC-like 1/SPARCL1
Applications Immunofluorescence
Classification Monoclonal
Clone 390
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias hevin, High endothelial venule protein, MAST 9, MAST9, PIG33, proliferation-inducing protein 33, SC1, SPARC-like 1 (hevin), SPARC-like 1 (mast9, hevin), SPARC-like protein 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human SPARC-like 1/SPARCL1 (NP_004675.3; Met1-Phe664).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8404
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.