missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPARC-like 1/SPARCL1 Antibody (204), mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
SPARC-like 1/SPARCL1 Monoclonal antibody specifically detects SPARC-like 1/SPARCL1 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | SPARC-like 1/SPARCL1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 204 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | hevin, High endothelial venule protein, MAST 9, MAST9, PIG33, proliferation-inducing protein 33, SC1, SPARC-like 1 (hevin), SPARC-like 1 (mast9, hevin), SPARC-like protein 1 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human SPARC-like 1/SPARCL1 (Accession#: NP_004675.3; Met 1-Phe 664). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?