Learn More
SPARC Antibody (001), Janelia Fluor™ 549, Novus Biologicals™
Product Code. 18427708 Shop All Bio Techne Products
Product Code. 18427708 Supplier Novus Biologicals Supplier No. NBP290612JF549
Rabbit Monoclonal Antibody
SPARC Monoclonal antibody specifically detects SPARC in Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation