missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPARC Antibody (001), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90612UV
This item is not returnable.
View return policy
Description
SPARC Monoclonal specifically detects SPARC in Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| SPARC | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, ELISA | |
| 1 | |
| Western Blot, ELISA | |
| Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin) | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SPARC (NP_033268.1; Met1-Ile302). | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Cell Cycle and Replication | |
| 6678 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction