missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPANXC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583819
This item is not returnable.
View return policy
Description
Immunogen sequence: SDPQPAPKKL KTSESSTILV VRYRRN Highest antigen sequence indentity to the following orthologs: Mouse - 42%, Rat - 42%.
Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family, which is located in a gene cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene encodes a protein that localizes to the nucleus and is expressed in highly metastatic cell lines, making the protein a potential diagnostic and prognostic marker. The protein belongs to a family of cancer/testis antigens and represents a potential target for cancer immunotherapy.
Specifications
| SPANXC | |
| Polyclonal | |
| Unconjugated | |
| SPANXC | |
| C; cancer/testis antigen 11.3; cancer/testis antigen family 11, member 3; cancer/testis-associated protein CTp11; cancer/testis-associated protein of 11 kD; cancer-testis-associated protein CTp11; CT11.3; CTp11; nuclear-associated protein SPAN-Xc; SPANX family member C; SPANX family, member C; SPANXC; SPANX-C; SPAN-Xc protein; Sperm protein associated with the nucleus on the X chromosome C; sperm protein associated with the nucleus, X chromosome, family member C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64663 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NY87 | |
| SPANXC | |
| Recombinant protein corresponding to Human SPANXC. Recombinant protein control fragment (Product #RP-99048). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction