missing translation for 'onlineSavingsMsg'
Learn More

SPANX-D Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£328.00 - £513.00

Specifications

Antigen SPANX-D
Dilution Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18613037
View Documents
Novus Biologicals
NBP2-68961-25ul
25 μL
£328.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18629176
View Documents
Novus Biologicals
NBP2-68961
100 μg
£513.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SPANX-D Polyclonal antibody specifically detects SPANX-D in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

SPANX-D
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
CT11.3;CT11.4;dJ171K16.1;SPANXC;SPANX-C;SPANXD;SPANX-D;SPANXE;SPANX-E;Sperm protein associated with the nucleus on the X chromosome D
This antibody was developed against a recombinant protein corresponding to amino acids: TSPEELVNDHARKNRINPLQMEEEE
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Polyclonal
Purified
RUO
PBS (pH 7.2) and 40% Glycerol
64648
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.