missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SPAM1 Synthetic Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
located within the following region: QQQNVQLSLTEATEKAKQEFEKAGKDFLVETIKLGKLLRPNHLWGYYLFP (Uniprot: P38567)
Specifications
Specifications
| Gene ID (Entrez) | 6677 |
| Purity | Multi-step |
| Concentration | LYOPH |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-83580. It may block above mentioned antibody from binding to its target protein in western blot and/or immunohistochemistry under proper experimental settings. |
| Gene Alias | EC 3.2.1.35, HYA1, HYAL1, HYAL3, HYAL5, Hyal-PH20, hyaluronidase PH-20, Hyaluronoglucosaminidase PH-20, MGC26532, PH20, PH-20, SPAG15, Sperm adhesion molecule 1, sperm adhesion molecule 1 (PH-20 hyaluronidase, zona pellucida binding), Sperm surface protein PH-20 |
| Gene Symbol | SPAM1 |
| Molecular Weight (g/mol) | M.W. Theoretical: 55 kDa |
| Quantity | 100 μg |
| Research Category | Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction