missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPAK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115976
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total STK39.
STK39 (STLK3) is involved in the cellular stress response pathway. STK39 is activated in response to hypotonic stress leading to phosphorylation of several cation-chloride-coupled co-transporters. STK39 activates the p38 MAP kinase pathway and its interaction with p38 decreases during cellular stress. STK39 acts as an intermediate in the response to cellular stress. STK39 is also an independent risk factor for hypertension in men and its intragenic SNPs can interact and function in the control of blood pressure.
Specifications
| SPAK | |
| Polyclonal | |
| Unconjugated | |
| STK39 | |
| AW227544; AW556857; DCHT; DKFZp686K05124; kidney-specific SPAK; pancreatic serine/threonine-protein kinase; Pask; proline-alanine-rich STE20-related kinase; PS/TK; PSTK1; RF005; Rnl5; serine threonine kinase 39; serine threonine kinase 39 (STE20/SPS1 homolog, yeast); serine/threonine kinase 39; serine/threonine kinase 39, STE20/SPS1 homolog; serine/threonine-protein kinase 39; small intestine SPAK-like kinase; Spak; Ste-20 related kinase; STE20/SPS1 homolog; STE20/SPS1-related proline-alanine-rich protein kinase; Ste20-like protein kinase; ste-20-related kinase; Stk39 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27347, 53416, 54348 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88506, Q9UEW8, Q9Z1W9 | |
| STK39 | |
| A synthesized peptide derived from human STK39(Accession Q9UEW8), corresponding to amino acid residues D381-V431. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction