missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Spaglumic Acid
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Purity | 98.0% |
| CAS | 3106-85-2 |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | Neuroscience-Neurodegeneration |
| Molecular Weight (g/mol) | 304.25 g/mol |
| Product Type | Natural Products |
| Quantity | 10 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 250 mg/mL (821.69 mM∣Need ultrasonic) |
| Synonym | N-Acetylaspartylglutamic acid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction