missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAG8 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00026206-B01P
This item is not returnable.
View return policy
Description
SPAG8 Polyclonal antibody specifically detects SPAG8 in Human samples. It is validated for Western Blot
Specifications
| SPAG8 | |
| Polyclonal | |
| Western Blot | |
| AAH19247.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 26206 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| BS-84, HSD-1MGC26201, hSMP-1, SMP1, SMP-1, SPAG3, sperm associated antigen 8, Sperm membrane protein 1, Sperm membrane protein BS-84, sperm-associated antigen 8 | |
| SPAG8 (AAH19247.1, 1 a.a. - 501 a.a.) full-length human protein. METNGSTEGSRSRSRSLDIQPSSEGLGPTSEPFPSSDDSPRSALAAATAAAAAAASAAAATAAFTTAKAAALSTKTPAPCSEFMEPSSDPSLLGEPCAGPGFTHNIAHGSLGFEPVYVSCIAQDTCTTTDHSSNPGPVPGSSSGPVLGSSSGAGHGSGSGSGPGCGSVPGSGSGPGPGSGPGSGPGHGSGSHPGPASGPGPDTGPDSELSPCIPPGFRNLVADRVLNYTSWSQHCPWEPQKQPPWEFLQVLEPGARGLWKPPDIKGKLMVCYETLPRGQCLLYNWEEERATNHLDQVPSMQDGSESFFFRHGHRGLLTMQLKSPMPSSTTQKDSYQPPGNVYWPLRGKREAMLEMLLQHQICKEVQAEQEPTRKLFEVESVTHHDYRMELAQAGTPAPTKPHDYRQEQPETFWIQRAPQLPTWWPLPTQVPAAEDYLTWKEWGFTGVQEVLSALLRATPGEYSVNICGMNEHPVCSRTWTNRLCHQEMGSKKTVTQEDRGW | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction